Specifications
Siedepunkt150°C (1013 mmHg)
CAS7758-99-8
Chemischer Name oder MaterialCopper(II) sulfate pentahydrate
FarbeBlau
Schmelzpunkt110°C
SummenformelCuSO4·5H2O
VerpackungNMA glass bottle, single shipper
SynonymCupric sulfate pentahydrate, Copper sulfate pentahydrate, Copper sulphate pentahydrate, Cupric sulphate pentahydrate
GüteMulti-Kompendium/USP
Molekulargewicht (g/mol)249.69
Reinheit (%)98.5 to 100.5%
Physikalische FormSolid
Menge500 g
pH3.5 to 4.5 (50 g/L, H2O, 20°C)
Unit Size500 g