3,4-Diaminobenzophenone, 99%
3,4-Diaminobenzophenone, 99%
3,4-Diaminobenzophenone, 99%
Thermo Scientific Chemicals

3,4-Diaminobenzophenone, 99%

CAS: 39070-63-8 | C13H12N2O | 212.25 g/mol
製品番号(カタログ番号)数量
18480025025 g
製品番号(カタログ番号) 184800250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS39070-63-8
IUPAC Name4-benzoylbenzene-1,2-diamine
Molecular FormulaC13H12N2O
InChI KeyRXCOGDYOZQGGMK-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1N)C(=O)C1=CC=CC=C1
さらに表示
Appearance (Color)Yellow to orange-brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point115°C to 119°C
Titration with HClO496.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only