Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Thermo Scientific Chemicals

Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 185002500
Price (USD)
110.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
110.00
Each
Chemical Identifiers
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay48 to 52 % Solids
Molecular weight3000 to 5000 (approx)
Appearance (Form)Clear to hazy liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only