Triisobutylaluminium, 1.0M solution in toluene, AcroSeal™
Triisobutylaluminium, 1.0M solution in toluene, AcroSeal™
Triisobutylaluminium, 1.0M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Triisobutylaluminium, 1.0M solution in toluene, AcroSeal™

CAS: 100-99-2 | C12H27Al | 198.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 185421000
Price (USD)
69.40
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
69.40
Each
Chemical Identifiers
CAS100-99-2
IUPAC Nametris(2-methylpropyl)alumane
Molecular FormulaC12H27Al
InChI KeyMCULRUJILOGHCJ-UHFFFAOYSA-N
SMILESCC(C)C[Al](CC(C)C)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric22.4 to 25.5 % (on Al basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only