BOC-ON, 99%
BOC-ON, 99%
BOC-ON, 99%
Thermo Scientific Chemicals

BOC-ON, 99%

CAS: 58632-95-4 | C13H14N2O3 | 246.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 185890050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS58632-95-4
IUPAC Nametert-butyl (Z)-[cyano(phenyl)methylidene]amino carbonate
Molecular FormulaC13H14N2O3
InChI KeyQQWYQAQQADNEIC-RVDMUPIBSA-N
SMILESCC(C)(C)OC(=O)O\N=C(/C#N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Melting point86°C to 89°C
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only