5-Nitro-1,10-fenantrolina, 98 %, Thermo Scientific Chemicals
5-Nitro-1,10-fenantrolina, 98 %, Thermo Scientific Chemicals
5-Nitro-1,10-fenantrolina, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Nitro-1,10-fenantrolina, 98 %, Thermo Scientific Chemicals

CAS: 4199-88-6 | C12H7N3O2 | 225.21 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 186080050
Precio (EUR)
252,00
Each
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
252,00
Each
Identificadores químicos
CAS4199-88-6
IUPAC Name5-nitro-1,10-phenanthroline
Molecular FormulaC12H7N3O2
InChI KeyPDDBTWXLNJNICS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC2=CC=CN=C2C2=NC=CC=C12
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)Light yellow to yellow-brown
Appearance (Form)Crystalline powder
Melting point199°C to 203°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only