Ethyl nipecotate, 97%
Ethyl nipecotate, 97%
Ethyl nipecotate, 97%
Thermo Scientific Chemicals

Ethyl nipecotate, 97%

CAS: 5006-62-2 | C8H15NO2 | 157.21 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 186620250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5006-62-2
IUPAC Nameethyl piperidine-3-carboxylate
Molecular FormulaC8H15NO2
InChI KeyXIWBSOUNZWSFKU-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4580 to 1.4610 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only