Bromotrimethylsilane, 98%
Bromotrimethylsilane, 98%
Bromotrimethylsilane, 98%
Thermo Scientific Chemicals

Bromotrimethylsilane, 98%

CAS: 2857-97-8 | C3H9BrSi | 153.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 186650050
Price (USD)
51.65
Online Exclusive
57.40
Save 5.75 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS2857-97-8
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4200 to 1.4250 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Silylation reagent
RUO – Research Use Only