3,4-Dimetoxifenol, 97 %, Thermo Scientific Chemicals
3,4-Dimetoxifenol, 97 %, Thermo Scientific Chemicals
3,4-Dimetoxifenol, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dimetoxifenol, 97 %, Thermo Scientific Chemicals

CAS: 2033-89-8 | C8H10O3 | 154.17 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
10 g
50 g
Envase:
Frasco de vidrio
Número de catálogo 186700100
Precio (USD)
-
Cantidad:
10 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS2033-89-8
IUPAC Name3,4-dimethoxyphenol
Molecular FormulaC8H10O3
InChI KeySMFFZOQLHYIRDA-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C=C1OC
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Off-white to light beige
Melting point77°C to 82°C
GC>=96.0 %
Appearance (Form)Crystalline powder
AppearanceMay darken on storage
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only