Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 186968000
Price (USD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13292-87-0
IUPAC Name(methylsulfanyl)methane borane
Molecular FormulaC2H9BS
InChI KeyRMHDLBZYPISZOI-UHFFFAOYSA-N
SMILESB.CSC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Strength1.90 to 2.20 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only