12-Crown-4, 97%
12-Crown-4, 97%
12-Crown-4, 97%
Thermo Scientific Chemicals

12-Crown-4, 97%

CAS: 294-93-9 | C8H16O4 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 187020025
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.60
Save 10.95
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.60
Save 10.95
Each
Chemical Identifiers
CAS294-93-9
IUPAC Name1,4,7,10-tetraoxacyclododecane
Molecular FormulaC8H16O4
InChI KeyXQQZRZQVBFHBHL-UHFFFAOYSA-N
SMILESC1COCCOCCOCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4600 to 1.4630 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only