Tetramethylammonium bromide, 98%
Tetramethylammonium bromide, 98%
Tetramethylammonium bromide, 98%
Thermo Scientific Chemicals

Tetramethylammonium bromide, 98%

CAS: 64-20-0 | C4H12BrN | 154.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 187631000
Price (USD)
55.65
Online exclusive
61.40
Save 5.75
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
55.65
Online exclusive
61.40
Save 5.75
Each
Chemical Identifiers
CAS64-20-0
IUPAC Nametetramethylazanium bromide
Molecular FormulaC4H12BrN
InChI KeyDDFYFBUWEBINLX-UHFFFAOYSA-M
SMILES[Br-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only