4-Amino-1-benzylpiperidine, 98%
4-Amino-1-benzylpiperidine, 98%
4-Amino-1-benzylpiperidine, 98%
Thermo Scientific Chemicals

4-Amino-1-benzylpiperidine, 98%

CAS: 50541-93-0 | C12H18N2 | 190.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 187660050
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.90
Save 5.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.90
Save 5.25
Each
Chemical Identifiers
CAS50541-93-0
IUPAC Name1-benzylpiperidin-4-amine
Molecular FormulaC12H18N2
InChI KeyYUBDLZGUSSWQSS-UHFFFAOYSA-N
SMILESNC1CCN(CC2=CC=CC=C2)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=97.5 %
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only