2-Thiopheneacetic acid, 98%
2-Thiopheneacetic acid, 98%
2-Thiopheneacetic acid, 98%
Thermo Scientific Chemicals

2-Thiopheneacetic acid, 98%

CAS: 1918-77-0 | C6H6O2S | 142.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 187730050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1918-77-0
IUPAC Name2-(thiophen-2-yl)acetic acid
Molecular FormulaC6H6O2S
InChI KeySMJRBWINMFUUDS-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point61°C to 66°C
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only