2,2-Dimethoxy-2-phenylacetophenone, 99%
2,2-Dimethoxy-2-phenylacetophenone, 99%
2,2-Dimethoxy-2-phenylacetophenone, 99%
Thermo Scientific Chemicals

2,2-Dimethoxy-2-phenylacetophenone, 99%

CAS: 24650-42-8 | C16H16O3 | 256.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 187840050
Price (USD)
41.65
Online exclusive
45.90
Save 4.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Online exclusive
45.90
Save 4.25
Each
Chemical Identifiers
CAS24650-42-8
IUPAC Name2,2-dimethoxy-1,2-diphenylethan-1-one
Molecular FormulaC16H16O3
InChI KeyKWVGIHKZDCUPEU-UHFFFAOYSA-N
SMILESCOC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point64°C to 68°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only