Ethoxycarbonyl isothiocyanate, 95%
Ethoxycarbonyl isothiocyanate, 95%
Ethoxycarbonyl isothiocyanate, 95%
Thermo Scientific Chemicals

Ethoxycarbonyl isothiocyanate, 95%

CAS: 16182-04-0 | C4H5NO2S | 131.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 187850250
Price (EUR)
203,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
203,00
Each
Chemical Identifiers
CAS16182-04-0
IUPAC Nameethyl N-carbothioylcarbamate
Molecular FormulaC4H5NO2S
InChI KeyBDTDECDAHYOJRO-UHFFFAOYSA-N
SMILESCCOC(=O)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to orange
Infrared spectrumConforms
GC>=94 %
Refractive index1.4980 to 1.5020 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only