Pentafluorobenzonitrile, 99%
Pentafluorobenzonitrile, 99%
Pentafluorobenzonitrile, 99%
Thermo Scientific Chemicals

Pentafluorobenzonitrile, 99%

CAS: 773-82-0 | C7F5N | 193.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 188341000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS773-82-0
IUPAC Namepentafluorobenzonitrile
Molecular FormulaC7F5N
InChI KeyYXWJGZQOGXGSSC-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C#N)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4410 to 1.4430 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only