4-Morpholinoaniline, 98+%
4-Morpholinoaniline, 98+%
4-Morpholinoaniline, 98+%
Thermo Scientific Chemicals

4-Morpholinoaniline, 98+%

CAS: 2524-67-6 | C10H14N2O | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 188650050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2524-67-6
IUPAC Name4-(morpholin-4-yl)aniline
Molecular FormulaC10H14N2O
InChI KeyPHNDZBFLOPIMSM-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)N1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.0 %
Melting point128°C to 132°C
Appearance (Color)Brown to purple
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only