3,4-Dimethoxyphenethyl alcohol, 98%, Thermo Scientific Chemicals
3,4-Dimethoxyphenethyl alcohol, 98%, Thermo Scientific Chemicals
3,4-Dimethoxyphenethyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dimethoxyphenethyl alcohol, 98%, Thermo Scientific Chemicals

CAS: 7417-21-2 | Molecular Formula: C10H14O3 | Molecular Weight: 182.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
18900010010 gGlass bottle
Catalog number 189000100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7417-21-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
GC>=97.5 %
Infrared spectrumConforms
Melting point44°C to 48°C
Appearance (Form)Powder and/or chunks

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only