Chest Freezer Racks
Actual product may vary
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Thermo Scientific™
Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)
189001General Freezer Items12.7 cu. ft. Freezers
114048General Freezer Items12.7, 20 cu. ft. Freezers
189000General Freezer Items20 cu. ft. Freezers
398184Boxes (2 in.)3 cu. ft. Chest Freezers
398185Boxes (3 in.)3 cu. ft. Freezers
398186Boxes (2 in.)13, 17, 20 cu. ft. Freezers
398187Boxes (3 in.)13, 17, 20 cu. ft. Freezers
398188Plates12.7, 17, 20 cu. ft. Freezers
Catalog number 189001
Price (MXN)
-
Holds:
General Freezer Items
For Use With (Equipment):
12.7 cu. ft. Freezers
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)12.7 cu. ft. Freezers
Height (Metric)30.2 cm
ShelvesNone
Width (Metric)14 cm
AutoclavableAutoclavable
ColorSilver
HoldsGeneral Freezer Items
MaterialStainless Steel
Positions12
Product TypeWire Basket (Rail Set)
ShapeRectangular
StackableYes
SterilityNon-sterile
Unit SizeEach