PBS Tablets
PBS Tablets
PBS Tablets
Gibco™

PBS Tablets

Phosphate-buffered saline (PBS) is a balanced salt formulation used for a variety of cell culture applications, such as washing cellsRead more
Have Questions?
Catalog NumberQuantity
18912014100 Tablets
Catalog number 18912014
Price (MXN)
8,748.57
Each
Quantity:
100 Tablets
Price (MXN)
8,748.57
Each

Phosphate-buffered saline (PBS) is a balanced salt formulation used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Each 5 g Gibco™ PBS Tablet should be dissolved in 500 mL of distilled water. The pH will be 7.45 and requires no adjustment.

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

These PBS Tablets are manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red


The complete formulation is available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPhosphate Buffered Saline (PBS)
IncludesNo Magnesium
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Recommended StorageStorage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Concentration1 X
Physical FormSolid
Product LineGibco
Quantity100 Tablets
pH7.45
Unit SizeEach

Frequently asked questions (FAQs)

Are your PBS products tested for endotoxin levels?

Our PBS, pH 7.2 and PBS, pH 7.4 products are tested for endotoxin and the endotoxin levels are shown on the Certificate of Analysis (CoA) for the particular lot number of the product.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products sterile and suitable for cell culture?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625), which is molecular biology grade and is certified to be RNase-free. However, it is not sterile and hence is not cell culture-grade.

All our other PBS products (Cat. Nos. 70011xxx, 10010xxx, 70013xxx, 20012xxx) are filter-sterilized using a 0.1 µm filter and are cell culture grade (ready to use directly on cells). However, they are not tested for nuclease activity.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products DNase- and RNase-free?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625) that is molecular biology grade and is certified to be RNase-free. Our other PBS products are made with distilled water and are filter-sterilized, but are not tested for RNase and DNase activity.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.