Specifications
Certifications/ComplianceCE Cat. III: EN 511, EN 388, EN ISO 21420
DescriptionMid-Arm Length
For Use With (Application)Cryogenic Preservation
Length (English)14 to 15 in.
Length (Metric)35.5 to 38 cm
Product TypeWaterproof Cryogenic Gloves
SizeSmall
Unit SizeEach