Tempshield™ Cryo Gloves - Elbow Length
Thermo Scientific™

Tempshield™ Cryo Gloves - Elbow Length

Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoSize
189446Large
189445Medium
189447X-Large
Número do catálogo 189446
Preço (BRL)
-
Size:
Large
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods. Safety certified to meet EU standards for CE Cat. III: EN511, EN388, EN ISO 21420. Available in wrist, elbow, mid-arm, shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
CE Cat. III: EN 511, EN 388, EN ISO 21420
Especificações
Certifications/ComplianceCE Cat. III: EN 511, EN 388, EN ISO 21420
DescriptionElbow Length
For Use With (Application)Cryogenic Preservation
Length (English)18 in.
Length (Metric)45.7 cm
Product TypeWaterproof Cryogenic Gloves
SizeLarge
Unit SizeEach