Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis
Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis
Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis
Thermo Scientific Chemicals

Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis

CAS: 35103-79-8 | CsH3O2 | 167.93 g/mol
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 189522500
Price (JPY)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS35103-79-8
IUPAC Namecaesium(1+) hydrate hydroxide
Molecular FormulaCsH3O2
InChI KeyABSOMGPQFXJESQ-UHFFFAOYSA-M
SMILESO.[OH-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<25 ppm
Appearance (Form)Crystalline powder
Titration with HCl>=96.5 %
Lithium (Li)=<1 ppm
Rubidium (Rb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only