Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Thermo Scientific Chemicals

Sodium hydride, 60% dispersion in mineral oil, in soluble bags

CAS: 7646-69-7 | HNa | 24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 189860010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7646-69-7
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Appearance (Form)Tacky powder
Assay57 to 63 % (NaH, Gasvolumetric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Reduction reagent
RUO – Research Use Only