L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
Thermo Scientific Chemicals

L(+)-Lactic acid, 90% solution in water

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Plastic drum
Catalog number 189870051
Price (JPY)
-
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS79-33-4
IUPAC Name(2S)-2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-REOHCLBHSA-N
SMILESC[C@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration excess NaOH>=88 %
Density(20°C) 1.2 to 1.21 g/ml
IdentificationPasses test
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only