Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Thermo Scientific Chemicals

Germanium dioxide, 99.999%, (trace metal basis), electronic grade

CAS: 1310-53-8 | GeO2 | 104.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 190000100
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Specifications
Assay Percent Range99.999%
CAS1310-53-8
Chemical Name or MaterialGermanium dioxide
ColorWhite
Density6.2300g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only