See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Incubators
▸
Incubator Accessories
▸
Relative Humidity System Option
Thermo Scientific™
Relative Humidity System Option
For Use With:
Environmental Chambers (3948 and 3907)
Have Questions?
Catalog Number
Installation
1900139
Factory Installed
Catalog number
1900139
Price (USD)
-
Request A Quote
Installation:
Factory Installed
Use Thermo Scientific™ Relative Humidity System Options for Environmental Chambers to address specific requirements to the relative humidity system.
Specifications
Description
Heatless Dryer Kit for sub-ambient chamber RH
For Use With (Equipment)
Environmental Chambers 3948/3907
Installation
Factory Installed
Lighting
LED
Type
Relative Humidity System