Chrome Azurol S
Chrome Azurol S
Chrome Azurol S
Thermo Scientific Chemicals

Chrome Azurol S

CAS: 1667-99-8 | C23H13Cl2Na3O9S | 605.277 g/mol
製品番号(カタログ番号)数量パッケージ
190051000100 gGlass bottle
製品番号(カタログ番号) 190051000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1667-99-8
IUPAC Nametrisodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichloro-3-sulfonatophenyl)methyl}-2-hydroxy-3-methylbenzoate
Molecular FormulaC23H13Cl2Na3O9S
InChI KeyFUIZKNBTOOKONL-DPSBJRLESA-K
SMILES[Na+].[Na+].[Na+].CC1=CC(=CC(C([O-])=O)=C1O)C(=C1\C=C(C)C(=O)C(=C1)C([O-])=O)\C1=C(Cl)C=CC(=C1Cl)S([O-])(=O)=O
さらに表示
Appearance (Form)Crystalline powder
UV Lambda max424 to 436 nm (c=0.03 g/l in H2O)
Appearance (Color)Dark brown to black
Infrared spectrumConforms
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only