Mercurochrome, 24-27% Mercury
Mercurochrome, 24-27% Mercury
Mercurochrome, 24-27% Mercury
Thermo Scientific Chemicals

Mercurochrome, 24-27% Mercury

CAS: 129-16-8 | C20H8Br2HgNa2O6 | 750.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 190111000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS129-16-8
IUPAC Namemerbromin
Molecular FormulaC20H9Br2HgNa2O6
InChI KeyBFMYDTVEBKDAKJ-UHFFFAOYNA-L
SMILESO.[Na+].[Na+].[O-]C1=C(Br)C=C2C(OC3=C(C=C(Br)C([O-])=C3[Hg])C22OC(=O)C3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening crystals or granules or scales
Assay24 to 27 % (Hg)
Appearance (Color)Green
Assay18 to 22.5 % (Br2)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only