alpha-Naphtholphthalein, pure, indicator
alpha-Naphtholphthalein, pure, indicator
alpha-Naphtholphthalein, pure, indicator
Thermo Scientific Chemicals

alpha-Naphtholphthalein, pure, indicator

CAS: 596-01-0 | C28H18O4 | 418.448 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 190120010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS596-01-0
IUPAC Name3,3-bis(4-hydroxynaphthalen-1-yl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H18O4
InChI KeyHQHBAGKIEAOSNM-UHFFFAOYSA-N
SMILESOC1=C2C=CC=CC2=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=C2C=CC=CC2=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<5 % (110°C, 1 h)
Specific absorbance A (1%/1cm)>=550 (at lamda max)
Appearance (Color)Beige to pink to brown
UV Lambda max648 to 654 nm (in 0.1N NaOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only