Victoria Blue B, pure
Victoria Blue B, pure
Victoria Blue B, pure
Thermo Scientific Chemicals

Victoria Blue B, pure

CAS: 2580-56-5 | C33H32ClN3 | 506.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 190190250
Price (USD)
47.65
Online exclusive
52.40
Save 4.75
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Online exclusive
52.40
Save 4.75
Each
Chemical Identifiers
CAS2580-56-5
IUPAC Namebis[4-(dimethylamino)phenyl][4-(phenylamino)naphthalen-1-yl]methylium chloride
Molecular FormulaC33H32ClN3
InChI KeyQHHVGBIWIQLWOJ-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=C2C=CC=CC2=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening crystalline powder
UV Lambda max599 to 605 nm (in ethanol)
Appearance (Color)Dark brown
UV spectrumComplies
Loss on drying=<5 % (60°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only