Sodium azide, 99%, extra pure
Sodium azide, 99%, extra pure
Sodium azide, 99%, extra pure
Thermo Scientific Chemicals

Sodium azide, 99%, extra pure

CAS: 26628-22-8 | N3Na | 65.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Catalog number 190380025
Price (USD)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS26628-22-8
IUPAC Namesodium azide
Molecular FormulaN3Na
InChI KeyPXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES[Na+].[N-]=[N+]=[N-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Insoluble matter=<0.05 % (in water)
Sodium carbonate (Na2CO3)=<0.15 %
Appearance (Color)White
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only