Molybdenum hexacarbonyl, 98%
Molybdenum hexacarbonyl, 98%
Molybdenum hexacarbonyl, 98%
Thermo Scientific Chemicals

Molybdenum hexacarbonyl, 98%

Molybdenum hexacarbonyl, 98%, C6O6Mo, CAS Number-13939-06-5, hexacarbonylmolybdenu, carbon monooxide-molybdenum 6:1, hexacarbonylmolybdenum, may contain up to cr 5g, hexakis carbon monoxide molybdenum, molybdenumhexacarbonyl, hexakis methanidylidyneoxidanium molybdenum, 50g, 100.0, 97.5, CHEBI:30508 | CAS: 13939-06-5 | C6O6Mo | 264 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 190390100
Price (USD)
114.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
114.00
Each
Chemical Identifiers
CAS13939-06-5
IUPAC Namehexakis(methanidylidyneoxidanium) molybdenum
Molecular FormulaC6MoO6
InChI KeyKMKBZNSIJQWHJA-UHFFFAOYSA-N
SMILES[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Elemental Analysis>=97.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only