CaptureSelect™ MultiSpecies Albumin Depletion Product, 50 mL - FAQs
CaptureSelect™ MultiSpecies Albumin Depletion Product, 50 mL - FAQs
View additional product information for CaptureSelect™ MultiSpecies Albumin Depletion Product - FAQs (191085305, 191085310, 191085350)
1 product FAQs found
Can I use the High Select Top14 Abundant Protein Depletion Midi Spin Columns to deplete proteins from rodent samples?
The High Select Top14 Abundant Protein Depletion Midi Spin Columns have only been validated for human samples. We do not recommend using them for protein depletion in rodent samples. For protein depletion in rodent samples, we recommend the following multi-species depletion resins:
  • CaptureSelect MultiSpecies Albumin Depletion Product (Cat. No. 191085305)
  • Pierce Albumin Serum Depletion Kits (Cat. No. 89875)
  • Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.