CaptureSelect™ IgG3 (Hu) Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgG3 (Hu) Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
19130401010 mL
1913040055 mL
19130405050 mL
Número do catálogo 191304010
Preço (BRL)
-
Quantity:
10 mL

CaptureSelect IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.

For Research Use Only. Not for use in diagnostic procedures.
Especificações
Quantity10 mL
Stationary PhaseAnti-IgG3
Binding Capacity>6 g/L Resin
FormResin Slurry
Particle Size35 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Conteúdo e armazenamento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light