Poly(ethylene glycol), average M.W. 200
Poly(ethylene glycol), average M.W. 200
Poly(ethylene glycol), average M.W. 200
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 200

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic bottle
Catalog number 192210010
Price (USD)
80.40
Each
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.40
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Hydroxyl value534 to 591 mg KOH/g
Refractive index1.4580 to 1.4610 (20°C, 589 nm)
Appearance (Form)Clear liquid
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only