Poly(ethylene glycol methyl ether), average M.W. 750
Poly(ethylene glycol methyl ether), average M.W. 750
Poly(ethylene glycol methyl ether), average M.W. 750
Thermo Scientific Chemicals

Poly(ethylene glycol methyl ether), average M.W. 750

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number 192320010
Price (TWD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9004-74-4
Molecular Formula(C2H4O)nCH4O
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
MDL NumberMFCD00084416
SpecificationsSpecification SheetSpecification Sheet
pH4.5 to 7.5 (5 % aq. soln., 25°C)
Infrared spectrumConforms
Appearance (Color)White to light yellow
Average molecular weight715 to 785
Appearance (Form)Solid or slightly hazy liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only