Lanthanum(III) oxide, 99.99%, (trace metal basis)
Lanthanum(III) oxide, 99.99%, (trace metal basis)
Lanthanum(III) oxide, 99.99%, (trace metal basis)
Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.99%, (trace metal basis)

CAS: 1312-81-8 | La2O3 | 325.82 g/mol
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 193295000
Price (JPY)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1312-81-8
IUPAC Namedilanthanum(3+) trioxidandiide
Molecular FormulaLa2O3
InChI KeyMRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Iron(III)oxide (Fe2O3)=<10 ppm
Appearance (Color)White
Appearance (Form)Powder
Assay>=99.94 % (La2O3) TREO base
Loss on ignition=<2.0 % (1000°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only