Magnesium oxide, 99.99%, (trace metal basis), extra pure
Magnesium oxide, 99.99%, (trace metal basis), extra pure
Magnesium oxide, 99.99%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Magnesium oxide, 99.99%, (trace metal basis), extra pure

CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 193430500
Price (USD)
257.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
257.00
Each
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total trace metal impurities=<0.01 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only