Potassium sulfate, pure
Potassium sulfate, pure
Potassium sulfate, pure
Thermo Scientific Chemicals

Potassium sulfate, pure

CAS: 7778-80-5 | K2O4S | 174.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 193810010
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<1 % (130°C)
pH5.5 to 8.5
Arsenic (As)=<0.0003 %
Selenium (Se)=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only