Sodium chloride, extra pure
Sodium chloride, extra pure
Sodium chloride, extra pure
Thermo Scientific Chemicals

Sodium chloride, extra pure

CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 194090250
Price (USD)
338.65
Special offer
Online exclusive
Ends: 28-Dec-2026
398.00
Save 59.35
Each
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (USD)
338.65
Special offer
Online exclusive
Ends: 28-Dec-2026
398.00
Save 59.35
Each
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<20 ppm
Sulfate (SO4)=<100 ppm
Barium (Ba)=<50 ppm
Calcium (Ca)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only