Sodium sulfate, 99+%, for HPLC, anhydrous
Sodium sulfate, 99+%, for HPLC, anhydrous
Sodium sulfate, 99+%, for HPLC, anhydrous
Thermo Scientific Chemicals

Sodium sulfate, 99+%, for HPLC, anhydrous

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 194100500
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.70
Save 8.05 (14%)
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.70
Save 8.05 (14%)
Each
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=99.0 %
Acidity/alkalinity=<0.0002 meq/g
UVat 220 nm A: =<0.1
UVat 250 nm A: =<0.02
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only