Donor Horse Serum, heat inactivated, New Zealand origin (custom product), 4 L - FAQs
Donor Horse Serum, heat inactivated, New Zealand origin (custom product), 4 L - FAQs
View additional product information for Donor Horse Serum, heat inactivated, New Zealand origin (custom product) - FAQs (1941300PP)
1 product FAQs found
When should I use heat-inactivated serum?
Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.