CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1943110055 mL
19431101010 mL
19431105050 mL
Catalog number 194311005
Price (USD)
1,250.00
Each
Quantity:
5 mL
Price (USD)
1,250.00
Each

CaptureSelect IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA. It shows no binding to other species IgA or other immunoglobulins.

This product can be used for purification of:

  • IgA1 and IgA2 from plasma and serum
  • Mono- and dimeric IgA, including secretory IgA
  • IgA Fab fragments
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity5 mL
Stationary PhaseAnti-IgA-CH1 (Hu)
Capacity (Metric)>6 g/L resin
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light