CaptureSelect™ FSH Affinity Matrix
Thermo Scientific™

CaptureSelect™ FSH Affinity Matrix

Thermo Scientific™ CaptureSelect™ FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
19431801L1 L
1943180055 mL
19431801010 mL
19431805050 mL
1943180250250 mL
Número do catálogo 19431801L
Preço (BRL)
244.375,87
Each
Quantity:
1 L
Preço (BRL)
244.375,87
Each

CaptureSelect FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Especificações
Flow Rate200 to 400 cm/hr.
MatrixAgarose-based, aldehyde-activated
Quantity1 L
Stationary PhaseAnti-Follicle Stimulating Hormone
Binding Capacity∼3 g FSH/L of Matrix
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Conteúdo e armazenamento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light