Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
19431805L | 5 L | Contact Us › | - | - | |
194318005 | 5 mL | 1,178.00 | - | ||
194318010 | 10 mL | 2,170.00 | - | ||
194318050 | 50 mL | 7,690.00 | - | ||
1943180250 | 250 mL | Contact Us › | - | - | |
19431801L | 1 L | Contact Us › | - | - |
CaptureSelect FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).