CaptureSelect™ LC-lambda (mouse) Affinity Matrix
CaptureSelect™ LC-lambda (mouse) Affinity Matrix
Thermo Scientific™
CaptureSelect™ LC-lambda (mouse) Affinity Matrix
Thermo Scientific™ CaptureSelect™ LC-lambda (mouse) Affinity Matrix contains a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) recognizing the lambda light chain of mouse Ig's.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19432305050 mL
1943230055 mL
19432301010 mL
Catalog number 194323050
Price (USD)
10,930.00
Each
Add to cart
Quantity:
50 mL
Price (USD)
10,930.00
Each
Add to cart
CaptureSelect LC-lambda (mouse) Affinity Matrix contains a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) recognizing the lambda light chain of mouse Ig's. The ligand is directed towards a unique domain on the constant part of the lambda light chain of immunoglobulins, which enables purification of mouse immunoglobulins and fab fragments without cross reactivity towards bovine immunoglobulins.This makes the CaptureSelect LC-lambda (mouse) Affinity Matrix a unique resin in terms of specificity.
The ligand is coupled on a 70 μm aldehyde-activated agarose bead. This matrix can be used for the purification and isolation of lambda light chain-containing mouse immunoglobulins and fab fragments from complex sources like cell culture supernatants, plasma, and serum in a single step. Due to its non-reactivity to bovine Ig, FCS can be applied in growth media without disturbing purification.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
MatrixAgarose-based, aldehyde-activated
Quantity50 mL
SpeciesMouse
Stationary PhaseAnti-LC-lambda
FormLiquid Suspension
Particle Size70 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.