CaptureSelect™ CH1-XL Affinity Matrix
Thermo Scientific™

CaptureSelect™ CH1-XL Affinity Matrix

Thermo Scientific™ CaptureSelect™ CH1-XL Affinity Matrix is the improved version of CaptureSelect IgG-CH1 Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
194346205050 mL
19434620055 mL
194346201010 mL
1943462250250 mL
194346201L1 L
194346205L5 L
194346210L10 L
Catalog number 1943462050
Price (USD)
6,980.00
Each
Quantity:
50 mL
Price (USD)
6,980.00
Each

CaptureSelect CH1-XL Affinity Matrix is the improved version of CaptureSelect IgG-CH1 Affinity Matrix. The specificity is the same, but it features a wider elution range (pH 3.0-4.5), increased binding capacity, and improved stability at higher pH.

CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material.

CaptureSelect CH1-XL Affinity Matrix features include:

  • Platform approach for purification of IgGs and their Fab fragments directly from complex source materials in a single step with high purity and yield
  • Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
  • No co-purification of overproduced free light chain due to selectivity for the CH1 domain
  • No binding to bovine antibodies, so presence of fetal bovine serum is not a problem
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs lower capacities can be observed.
  • Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
  • Flow rate: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeCH1-XL affinity ligand
MatrixAgarose-based, epoxide-activated
Quantity50 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Stationary PhaseAnti-IgG-CH1
Binding Capacity>15 mg/mL
FormAgarose Bead
Particle Size65 μm
Product LineCaptureSelect
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.