Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis)
Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis)
Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis)

CAS: 10035-01-5 | Cl3Yb·6H2O | 387.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 194430500
Price (USD)
364.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
364.00
Each
Chemical Identifiers
CAS10035-01-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Ytterbium (Yb)43.1 to 46.2 %
Total conc. of imp.as REO=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only